Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3-Hydroxyphenyl)-DL-alanine, 98%

Chemical Identifiers

Chemical Name or Material 3-(3-Hydroxyphenyl)-DL-alanine
CAS 775-06-4
Formula Weight 181.19
MDL Number MFCD00002597
Beilstein 2416853
Merck Index 14,9840
Synonym DL-m-Tyrosine; (±)-2-Amino-3-(3-hydroxyphenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6316303

  SDS
alfa aesar™
H6316303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6316306

  SDS
alfa aesar™
H6316306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63163MD

  SDS
alfa aesar™
H63163MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.19
Melting Point 280° to 285°C (dec.)
Chemical Name or Material 3-(3-Hydroxyphenyl)-DL-alanine
Physical Form Powder
Assay 98%
MDL Number MFCD00002597
CAS 775-06-4
Beilstein 2416853
Merck Index 14,9840
Synonym DL-m-Tyrosine; (±)-2-Amino-3-(3-hydroxyphenyl)propionic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color Off-white to yellow
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number YP2278000

Applications
3-(3-Hydroxyphenyl)-DL-alanine is used as a food and feed additive. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 1M HCl.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.