Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3-Fluoro-4-methoxybenzoyl)propionic acid, 99%

$67.11 - $224.73

Chemical Identifiers

Chemical Name or Material 3-(3-Fluoro-4-methoxybenzoyl)propionic acid
CAS 347-63-7
Molecular Formula C11H11FO4
Formula Weight 226.2
MDL Number MFCD00020538
Beilstein 3322421
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0704203

 
alfa aesar™
L0704203
1g Each for $67.11

AAL0704206

 
alfa aesar™
L0704206
5g Each for $224.73
Description & Specifications

Specifications

Formula Weight 226.2
MDL Number MFCD00020538
CAS 347-63-7
Chemical Name or Material 3-(3-Fluoro-4-methoxybenzoyl)propionic acid
Assay 99%
Molecular Formula C11H11FO4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 3322421
Melting Point 169° to 173°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.