Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine, 97%

$111.95 - $382.72

Chemical Identifiers

Chemical Name or Material 3-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine
CAS 139269-06-0
Molecular Formula C9H15N3O
Formula Weight 181.24
MDL Number MFCD08061044
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5087903

  SDS
alfa aesar™
H5087903
1g Each for $382.72

AAH50879MD

  SDS
alfa aesar™
H50879MD
250mg Each for $111.95
Description & Specifications

Specifications

Formula Weight 181.24
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C9H15N3O
CAS 139269-06-0
MDL Number MFCD08061044
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-(3-Ethyl-1,2,4-oxadiazol-5-yl)piperidine
Quantity 1g
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.