Please login to your online account to display your discounted pricing

3,3-Diphenylpropylamine 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  159481000

 View more versions of this product

Catalog No. AC159481000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid After Melting
Formula Weight 211.31
Quantity 100g
Color Clear yellow
EINECS Number 226-984-3
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 3, 3-Diphenylpropylamine
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Refractive Index 1.5820 to 1.5840 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C15H17N
Linear Formula (C6H5)2CHCH2CH2NH2
MDL Number MFCD00008202
CAS 5586-73-2
CAS Min % 97.5

Specifications

Chemical Name or Material 3, 3-Diphenylpropylamine
Name Note 98%
CAS 5586-73-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15H17N
Linear Formula (C6H5)2CHCH2CH2NH2
Formula Weight 211.31
MDL Number MFCD00008202