Please login to your online account to display your discounted pricing

3,3-Diphenylpropylamine 98%, ACROS Organics™

$103.17 - $103.17

Chemical Identifiers

Chemical Name or Material 3, 3-Diphenylpropylamine
Name Note 98%
CAS 5586-73-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15H17N
Linear Formula (C6H5)2CHCH2CH2NH2
Formula Weight 211.31
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC159480250

  SDS
thermo fisher scientific
159480250
Glass bottle 25g Each for $103.17

AC159481000

  SDS
thermo fisher scientific
159481000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid After Melting
Assay ≥97.5%
Formula Weight 211.31
Quantity 25g
Color Clear yellow
EINECS Number 226-984-3
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 3, 3-Diphenylpropylamine
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Refractive Index 1.5820 to 1.5840 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C15H17N
Linear Formula (C6H5)2CHCH2CH2NH2
MDL Number MFCD00008202
CAS 5586-73-2
CAS Min % 97.5