Please login to your online account to display your discounted pricing

3,3-Diphenylhexamethyltrisiloxane, 97%, Acros Organics


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  336660050

 View more versions of this product

Catalog No. AC336660050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 360.68
Quantity 5g
Color Clear colorless
TSCA TSCA
Density 0.9800g/mL
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C18H28O2Si3
Linear Formula CH3Si(C6H5)2OSi(CH3)2OSi(CH3)3
Refractive Index 1.4900 to 1.4930 (20°C, 589nm)
CAS 797-77-3
Chemical Name or Material 3, 3-Diphenylhexamethyltrisiloxane
Name Note 97%
Assay Percent Range 97%
MDL Number MFCD00039794

Specifications

Chemical Name or Material 3, 3-Diphenylhexamethyltrisiloxane
Name Note 97%
CAS 797-77-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C18H28O2Si3
Linear Formula CH3Si(C6H5)2OSi(CH3)2OSi(CH3)3
Formula Weight 360.68
MDL Number MFCD00039794