Please login to your online account to display your discounted pricing

3,3-Diphenylhexamethyltrisiloxane, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 3, 3-Diphenylhexamethyltrisiloxane
Name Note 97%
CAS 797-77-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C18H28O2Si3
Linear Formula CH3Si(C6H5)2OSi(CH3)2OSi(CH3)3
Formula Weight 360.68
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC336660050

  SDS
thermo fisher scientific
336660050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 360.68
Quantity 5g
Color Clear colorless
TSCA TSCA
Density 0.9800g/mL
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C18H28O2Si3
Linear Formula CH3Si(C6H5)2OSi(CH3)2OSi(CH3)3
Refractive Index 1.4900 to 1.4930 (20°C, 589nm)
CAS 797-77-3
Chemical Name or Material 3, 3-Diphenylhexamethyltrisiloxane
Name Note 97%
Assay Percent Range 97%
MDL Number MFCD00039794