Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3'-Dinitrobenzophenone, 96%

$61.03 - $61.03

Chemical Identifiers

Chemical Name or Material 3,3'-Dinitrobenzophenone
CAS 21222-05-9
Molecular Formula C13H8N2O5
Formula Weight 272.22
MDL Number MFCD00039740
Beilstein 1888002
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2115306

  SDS
alfa aesar™
B2115306
5g Each for $61.03
Description & Specifications

Specifications

Formula Weight 272.22
MDL Number MFCD00039740
CAS 21222-05-9
Chemical Name or Material 3,3'-Dinitrobenzophenone
Molecular Formula C13H8N2O5
Health Hazard 1 Warning
Assay 96%
Melting Point 153° to 155°C
Beilstein 1888002
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.