Learn More
3,3-Dimethylpiperidine, 95%, Thermo Scientific Chemicals
$332.37 - $835.18
Chemical Identifiers
CAS | 1193-12-0 |
---|---|
Molecular Formula | C7H15N |
Molecular Weight (g/mol) | 113.20 |
MDL Number | MFCD00005995 |
InChI Key | CDODDZJCEADUQQ-UHFFFAOYSA-N |
PubChem CID | 70942 |
IUPAC Name | 3,3-dimethylpiperidine |
SMILES | CC1(C)CCCNC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH25753MD
|
Thermo Scientific Chemicals
H25753MD |
250 mg |
Each for $332.37
|
|
|||||
AAH2575303
|
Thermo Scientific Chemicals
H2575303 |
1 g |
Each for $835.18
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1193-12-0 | |
113.20 | |
CDODDZJCEADUQQ-UHFFFAOYSA-N | |
3,3-dimethylpiperidine |
C7H15N | |
MFCD00005995 | |
70942 | |
CC1(C)CCCNC1 |
Specifications
1193-12-0 | |
134°C to 136°C | |
Amine-like | |
1.4475 | |
250 mg | |
Air Sensitive | |
CDODDZJCEADUQQ-UHFFFAOYSA-N | |
3,3-dimethylpiperidine | |
70942 | |
95% |
0.81 | |
26°C (79°F) | |
C7H15N | |
MFCD00005995 | |
UN2733 | |
piperidine, 3,3-dimethyl, 3,3-dimethyl-piperidine, 3,3-dimethyl piperidine, pubchem10316, acmc-1bo7w, 3,3-dimethylpiperdine | |
CC1(C)CCCNC1 | |
113.20 | |
113.2 | |
3,3-Dimethylpiperidine |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 214-767-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only