Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3-Dimethylpentane, 99%

Chemical Identifiers

Chemical Name or Material 3,3-Dimethylpentane
CAS 562-49-2
Molecular Formula C7H16
Formula Weight 100.21
MDL Number MFCD00009322
UN Number UN1206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4370603

  SDS
alfa aesar™
4370603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4370606

  SDS
alfa aesar™
4370606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.21
Chemical Name or Material 3,3-Dimethylpentane
TSCA TSCA
Physical Form Liquid
Assay 99%
Molecular Formula C7H16
MDL Number MFCD00009322
CAS 562-49-2
UN Number UN1206
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEPTANES
Density 0.69g/mL
Flash Point −6°C (21°F)
Boiling Point 86°C
Melting Point −135°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.