Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3-Dimethyloxetane, 98%

Chemical Identifiers

Chemical Name or Material 3,3-Dimethyloxetane
CAS 6921-35-3
Molecular Formula C5H10O
Formula Weight 86.13g/mol
MDL Number MFCD00005168
UN Number UN3271
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27370AA

 
alfa aesar™
H27370-AA
5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 81°C
Quantity 5mL
Flash Point −9°C (16°F)
Chemical Name or Material 3,3-Dimethyloxetane
CAS 6921-35-3
Assay 98%
Molecular Formula C5H10O
Formula Weight 86.13g/mol
MDL Number MFCD00005168
UN Number UN3271
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.