Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3-Dimethylglutarimide, 99%

$53.88 - $126.68

Chemical Identifiers

Chemical Name or Material 3,3-Dimethylglutarimide
CAS 1123-40-6
Molecular Formula C7H11NO2
Formula Weight 141.17
MDL Number MFCD00006671
Beilstein 111981
Synonym 4,4-Dimethylpiperidine-2,6-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2219606

 
alfa aesar™
B2219606
5g Each for $53.88

AAB2219614

 
alfa aesar™
B2219614
25g Each for $126.68
Description & Specifications

Specifications

Formula Weight 141.17
Chemical Name or Material 3,3-Dimethylglutarimide
Assay 99%
Molecular Formula C7H11NO2
MDL Number MFCD00006671
CAS 1123-40-6
Beilstein 111981
Synonym 4,4-Dimethylpiperidine-2,6-dione
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number MA4348000
Melting Point 144° to 146°C
Quantity 5g

Applications
3,3′-dimethylglutarimide were used as the barbital subsitutes. 3,3-di­methyl­glutarimide is involved formation of the heterodimer.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.