Please login to your online account to display your discounted pricing

3,3-Dimethylglutaric Anhydride 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  116261000

 View more versions of this product

Catalog No. AC116261000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 142.15
Quantity 100g
Color White to off-white
Beilstein 17, 419
EINECS Number 223-998-1
TSCA TSCA
Melting Point 122° to 126°C
CAS Max % 100.0
Chemical Name or Material 3, 3-Dimethylglutaric anhydride
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
MDL Number MFCD00006684
CAS Min % 98.5
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99%
Molecular Formula C7H10O3
CAS 4160-82-1

Specifications

Chemical Name or Material 3, 3-Dimethylglutaric anhydride
Name Note 99%
CAS 4160-82-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H10O3
Formula Weight 142.15
MDL Number MFCD00006684
Accessories

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents