Learn More
3,3-Dimethylglutaric acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2038830
Description
It acts as a reactant involved in cyclodehydration of diols, synthesis of conjugates of betulin derivatives used as anti-HIV agents, preparation of dimeric peptide antagonists of IgG-FcRn interaction, microwave-assisted protection of glutaraldehyde, synthesis of glycyrrhetinic acid derivatives for proteasome inhibition and catalytic, asymmetric transannular aldolizations. It is also used with other dibasic esters as solvents in paints, cosmetics, and fragrances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt acts as a reactant involved in cyclodehydration of diols, synthesis of conjugates of betulin derivatives used as anti-HIV agents, preparation of dimeric peptide antagonists of IgG-FcRn interaction, microwave-assisted protection of glutaraldehyde, synthesis of glycyrrhetinic acid derivatives for proteasome inhibition and catalytic, asymmetric transannular aldolizations. It is also used with other dibasic esters as solvents in paints, cosmetics, and fragrances.
Solubility
Solubility in water, very faint turbidity.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
3,3-Dimethylglutaric acid | |
98°C to 103°C | |
89°C to 90°C (2 mmHg) | |
MFCD00002716 | |
1765439 | |
DUHQIGLHYXLKAE-UHFFFAOYSA-N | |
3,3-dimethylpentanedioic acid | |
20984 | |
160.17 |
4839-46-7 | |
1.42 | |
C7H12O4 | |
250 g | |
3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid | |
CC(C)(CC(O)=O)CC(O)=O | |
160.17 | |
CHEBI:68502 | |
≥98% |
Chemical Identifiers
4839-46-7 | |
160.17 | |
DUHQIGLHYXLKAE-UHFFFAOYSA-N | |
20984 | |
3,3-dimethylpentanedioic acid |
C7H12O4 | |
MFCD00002716 | |
3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid | |
CHEBI:68502 | |
CC(C)(CC(O)=O)CC(O)=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 225-425-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only