missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3-Dimethylbutyraldehyde, 97%

Manufacturer:  Alfa Aesar™ H55522AC

 View more versions of this product

Catalog No. AAH55522AC



Specifications

Specifications

3,3-Dimethylbutyraldehyde
13°C (55°F)
Air and moisture sensitive
25mL
C6H12O
MFCD00042807
1735829
LTNUSYNQZJZUSY-UHFFFAOYSA-N
3,3-dimethylbutanal
76335
97%
0.798
1.397
104°C to 106°C
2987-16-8
(CH3)3CCH2CHO
UN1989
3,3-dimethyl-butanal
CC(C)(C)CC=O
100.161
100.16

Chemical Identifiers

2987-16-8
100.161
LTNUSYNQZJZUSY-UHFFFAOYSA-N
76335
CC(C)(C)CC=O
C6H12O
MFCD00042807
3,3-dimethyl-butanal
3,3-dimethylbutanal
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 221-054-3

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents