missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3-Dimethylbutan-2-one oxime, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438310010

 View more versions of this product

Catalog No. AC438310010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3, 3-Dimethylbutan-2-one oxime
Glass bottle
White
1g
97.5
97.5% min. (GC)
2-butanone,3-dimethyl-, oxime, 2-hydroxyimino-3,3-dimethylbutane, 2e-n-hydroxy-3,3-dimethylbutan-2-imine, 3,3-dimethyl-2-butanone oxime, pinacolone oxime, pinakolinoxim
CC(=NO)C(C)(C)C
115.18
115.18
98%
Authentic
171°C to 172°C (748.0 Torr)
75°C to 77°C
2475-93-6
100.0
C6H13NO
UNSDDJQNHCSVSW-FNORWQNLSA-N
(NE)-N-(3,3-dimethylbutan-2-ylidene)hydroxylamine
9561079
Crystalline Powder

Chemical Identifiers

2475-93-6
115.18
2-butanone,3-dimethyl-, oxime, 2-hydroxyimino-3,3-dimethylbutane, 2e-n-hydroxy-3,3-dimethylbutan-2-imine, 3,3-dimethyl-2-butanone oxime, pinacolone oxime, pinakolinoxim
(NE)-N-(3,3-dimethylbutan-2-ylidene)hydroxylamine
C6H13NO
UNSDDJQNHCSVSW-FNORWQNLSA-N
9561079
CC(=NO)C(C)(C)C
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Contaminated work clothing should not be allowed out of the workplace.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If skin irritation or rash occurs: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents