Please login to your online account to display your discounted pricing

Alfa Aesar™ (3,3-Dimethylallyl)triphenylphosphonium bromide, 98+%


Quantity 25g

Manufacturer: alfa aesar™  L0051314

 View more versions of this product

Catalog No. AAL0051314

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 411.33
Molecular Formula C23H24BrP
Chemical Name or Material (3,3-Dimethylallyl)triphenylphosphonium bromide
CAS 1530-34-3
Synonym (3-Methyl-2-butenyl)triphenylphosphonium bromide; Prenyltriphenylphosphonium bromide
Assay ≥98%
MDL Number MFCD00051864
Beilstein 3641199
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 235° to 239°C
Quantity 25g
Health Hazard 1 Warning

Specifications

Chemical Name or Material (3,3-Dimethylallyl)triphenylphosphonium bromide
CAS 1530-34-3
Molecular Formula C23H24BrP
Formula Weight 411.33
MDL Number MFCD00051864
Beilstein 3641199
Synonym (3-Methyl-2-butenyl)triphenylphosphonium bromide; Prenyltriphenylphosphonium bromide
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents