Learn More
3,3-Dimethylacryloyl chloride, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 179730050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3, 3-Dimethylacryloyl chloride | |
3350-78-5 | |
100.0 | |
145.0°C to 147.0°C | |
Authentic | |
Glass bottle | |
1.4750 to 1.4780 | |
MFCD00000730 | |
02, 433 | |
3,3-dimethylacryloyl chloride, 3-methylcrotonoyl chloride, 3-methyl-but-2-enoyl chloride, 3,3-dimethylacrylyl chloride, 2-butenoyl chloride, 3-methyl, 3-methyl-2-butenoyl chloride, 3,3-dimethylacryloylchloride, dimethylacrylyl chloride, dimethyl acryloyl chloride, acmc-1cpb0 | |
BDUBTLFQHNYXPC-UHFFFAOYSA-N | |
3-methylbut-2-enoyl chloride | |
102394 | |
97% |
97% | |
96.0 | |
1.0600g/mL | |
51°C | |
96% min. (GC) | |
C5H7ClO | |
(CH3)2C=CHCOCl | |
5g | |
1.06 | |
Solubility in water: reacts | |
CC(C)=CC(Cl)=O | |
118.56 | |
118.56 |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 222-109-4
TSCA : TSCA
Recommended Storage : Product may darken on storage
RUO â Research Use Only