missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-3,3-Dimethyl-2-butylamine, 98%

Manufacturer:  Alfa Aesar™ A1153006

 View more versions of this product

Catalog No. AAA1153006


 

Description

Description

Applications
(+/-)-3,3-Dimethyl-2-butylamine is used to produce 1-pyridin-3-yl-3-(1,2,2-trimethyl-propyl)-thiourea. It is also used as pharmaceutical intermediates.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from acid chlorides and acid anhydrides.
Specifications

Specifications

(±)-3,3-Dimethyl-2-butylamine
98%
MFCD00008078
5g
DXSUORGKJZADET-UHFFFAOYSA-N
3,3-dimethylbutan-2-amine
520907
3850-30-4
C6H15N
UN2733
2-Amino-3,3-dimethylbutane
CC(C(C)(C)C)N
101.193
101.19

Chemical Identifiers

3850-30-4
101.193
DXSUORGKJZADET-UHFFFAOYSA-N
520907
C6H15N
MFCD00008078
2-Amino-3,3-dimethylbutane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

TSCA : TSCA

SDS Documents