Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3-Dimethyl-2-butanol, 99%

Chemical Identifiers

Chemical Name or Material 3,3-Dimethyl-2-butanol
CAS 464-07-3
Molecular Formula (CH3)3CCH(OH)CH3
Formula Weight 102.18
MDL Number MFCD00004522
UN Number UN1987
Synonym tert-Butyl methyl carbinol; Pinacolyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4372550

 
alfa aesar™
4372550
5 x 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 102.18
Chemical Name or Material 3,3-Dimethyl-2-butanol
TSCA TSCA
Physical Form Liquid
Assay 99%
Molecular Formula (CH3)3CCH(OH)CH3
MDL Number MFCD00004522
CAS 464-07-3
UN Number UN1987
Synonym tert-Butyl methyl carbinol; Pinacolyl alcohol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
RTECS Number EL2276000
Refractive Index 1.4150
Density 0.812g/mL
Flash Point 28°C (84°F)
Boiling Point 119° to 121°C
Melting Point 5.3°C
Quantity 5 x 10g
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.