Please login to your online account to display your discounted pricing

3,3-Dimethyl-1-butyne, 98%, ACROS Organics™

$74.71 - $74.71

Chemical Identifiers

Chemical Name or Material 3, 3-Dimethyl-1-butyne
CAS 917-92-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H10
Linear Formula (CH3)3CCidentical toCH
Formula Weight 82.13
MDL Number MFCD00008852
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC223440250

  SDS
thermo fisher scientific
AC223440250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC223440050

  SDS
thermo fisher scientific
223440050
Glass bottle 5mL Each for $74.71
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 82.13
Quantity 25mL
Color Clear colorless
Density 0.6600g/mL
Beilstein 01, 256
CAS Max % 100.0
Linear Formula (CH3)3CCidentical toCH
Synonym tert-Butylacetylene
Molecular Formula C6H10
Chemical Name or Material 3, 3-Dimethyl-1-butyne
Assay Percent Range 98%
MDL Number MFCD00008852
CAS 917-92-0
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
CAS Min % 97.5
Health Hazard 1 Danger
Packaging Glass bottle
Refractive Index 1.3730 to 1.3750 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.