Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3-Dimethyl-1-butene, 95%

$31.09 - $31.09

Chemical Identifiers

Chemical Name or Material 3,3-Dimethyl-1-butene
CAS 558-37-2
Molecular Formula C6H12
Formula Weight 84.16
MDL Number MFCD00008853
UN Number UN3295
Beilstein 1697101
Synonym tert-Butylethylene; Neohexene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL08693AD

 
alfa aesar™
L08693AD*
50mL Each for $31.09
Description & Specifications

Specifications

Formula Weight 84.16
Chemical Name or Material 3,3-Dimethyl-1-butene
TSCA TSCA
Assay 95%
Molecular Formula C6H12
MDL Number MFCD00008853
CAS 558-37-2
UN Number UN3295
Beilstein 1697101
Synonym tert-Butylethylene; Neohexene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Flash Point −28°C (−18°F)
Boiling Point 40° to 41°C
Melting Point −115°C
Quantity 50mL

Applications
It is used in the reaction pathways for the conversion of methanol and olefins on H-ZSM-5 zeolite.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

2° to 8°C