Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3-Dimethyl-1,2-butanediol, tech. 90%

$43.84 - $170.27

Chemical Identifiers

Chemical Name or Material 3,3-Dimethyl-1,2-butanediol
CAS 59562-82-2
Purity Grade Technical Grade
Molecular Formula C6H14O2
Formula Weight 118.18
MDL Number MFCD00010735
Beilstein 1732476
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0761606

  SDS
alfa aesar™
L0761606
5g Each for $43.84

AAL0761614

  SDS
alfa aesar™
L0761614
25g Each for $170.27
Description & Specifications

Specifications

Formula Weight 118.18
Chemical Name or Material 3,3-Dimethyl-1,2-butanediol
Molecular Formula C6H14O2
MDL Number MFCD00010735
CAS 59562-82-2
Assay 90%
Purity Grade Technical Grade
Boiling Point 201° to 202°C
Beilstein 1732476
Quantity 5g
Flash Point 98°C (208°F)
Melting Point 37° to 39°C