missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,3'-Dimethoxybenzil, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1156403
Description
3,3'-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,3′-Dimethoxybenzil used in the preparation of benzil derivatives. It is also used as an intermediate in organic synthesis and pharmaceuticals.
Solubility
Insoluble in water.
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
3,3'-Dimethoxybenzil | |
81°C to 83°C | |
MFCD00038221 | |
2382124 | |
Insoluble in water. | |
COC1=CC=CC(=C1)C(=O)C(=O)C2=CC(=CC=C2)OC | |
270.284 | |
270.29 |
40101-17-5 | |
C16H14O4 | |
1 g | |
3,3'-dimethoxybenzil, 1,2-bis 3-methoxyphenyl ethane-1,2-dione, ethanedione, bis 3-methoxyphenyl, 1,2-ethanedione, 1,2-bis 3-methoxyphenyl, bis 3-methoxyphenyl ethane-1,2-dione, bis 3-methoxyphenyl ethanedione, acmc-20a2cz, 3,3'-dimethoxy benzil, d01vau, benzil-based compound, 22 | |
PJGXOGKIVAJFTE-UHFFFAOYSA-N | |
1,2-bis(3-methoxyphenyl)ethane-1,2-dione | |
123493 | |
≥99% |
Chemical Identifiers
40101-17-5 | |
270.284 | |
PJGXOGKIVAJFTE-UHFFFAOYSA-N | |
123493 | |
COC1=CC=CC(=C1)C(=O)C(=O)C2=CC(=CC=C2)OC |
C16H14O4 | |
MFCD00038221 | |
3,3'-dimethoxybenzil, 1,2-bis 3-methoxyphenyl ethane-1,2-dione, ethanedione, bis 3-methoxyphenyl, 1,2-ethanedione, 1,2-bis 3-methoxyphenyl, bis 3-methoxyphenyl ethane-1,2-dione, bis 3-methoxyphenyl ethanedione, acmc-20a2cz, 3,3'-dimethoxy benzil, d01vau, benzil-based compound, 22 | |
1,2-bis(3-methoxyphenyl)ethane-1,2-dione |
Safety and Handling
EINECSNumber : 254-793-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only