Please login to your online account to display your discounted pricing

3,3'-Dimethoxybenzidine dihydrochloride, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  184470250

 View more versions of this product

Catalog No. AC184470250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.5% (on dry substance)
Formula Weight 317.21
Quantity 25g
Color Light beige to purple-gray
Beilstein 13, 807
EINECS Number 243-737-5
TSCA TSCA
RTECS Number DD1050000
CAS Max % 100.0
Chemical Name or Material 3, 3'-Dimethoxybenzidine dihydrochloride
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C14H16N2O2·2HCl
MDL Number MFCD00012488
CAS 20325-40-0
Synonym o-Dianisidine dihydrochloride
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Loss on Drying 1% max.

Specifications

Chemical Name or Material 3, 3'-Dimethoxybenzidine dihydrochloride
Name Note 99%
CAS 20325-40-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (on dry substance)
Assay Percent Range 99%
Molecular Formula C14H16N2O2·2HCl
Formula Weight 317.21
MDL Number MFCD00012488
Synonym o-Dianisidine dihydrochloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause cancer.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents