Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3-Difluoropiperidine hydrochloride, 99%

$187.80 - $554.14

Chemical Identifiers

Chemical Name or Material 3,3-Difluoropiperidine hydrochloride
CAS 496807-97-7
Molecular Formula C5H9F2N·HCl
Formula Weight 157.59
MDL Number MFCD03452800
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1977703

  SDS
alfa aesar™
L1977703
1g Each for $554.14

AAL19777MD

  SDS
alfa aesar™
L19777MD
250mg Each for $187.80
Description & Specifications

Specifications

Formula Weight 157.59
Chemical Name or Material 3,3-Difluoropiperidine hydrochloride
Assay 99%
MDL Number MFCD03452800
CAS 496807-97-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C5H9F2N·HCl
Melting Point ≈240°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.