Learn More
3,3'-Diethylthiadicarbocyanine iodide, Thermo Scientific Chemicals
As a solvent soluble laser dye
Supplier: Thermo Scientific Chemicals 04385109
Description
It is employed as a solvent soluble laser dye. DTDCi is used as fluorescent contrast agent in three-dimensional fluorescence lifetime tomography and as a dye for rapid genetic screening. It is a potential sensitive probe useful for fluorometric analysis of transferable membrane pores and effects of membrane potential on Na cotransports in eel intestinal brush-border membrane vesicles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3,3'-Diethylthiadicarbocyanine iodide | |
248°C (decomposition) | |
C23H24IN2S2+ | |
10 g | |
3838938 | |
3,3'-diethylthiadicarbocyanine iodide | |
MNQDKWZEUULFPX-UHFFFAOYSA-N | |
(2E)-3-ethyl-2-[(2E,4E)-5-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzothiazole;hydroiodide | |
91869351 | |
99% |
514-73-8 | |
Green | |
MFCD00074829 | |
UN2811 | |
14,3369 | |
Soluble in methanol | |
CCN1C2=CC=CC=C2SC1=CC=CC=CC3=[N+](C4=CC=CC=C4S3)CC.I | |
519.483 | |
518.47 | |
Powder |
Chemical Identifiers
514-73-8 | |
519.483 | |
MNQDKWZEUULFPX-UHFFFAOYSA-N | |
91869351 | |
CCN1C2=CC=CC=C2SC1=CC=CC=CC3=[N+](C4=CC=CC=C4S3)CC.I |
C23H24IN2S2+ | |
MFCD00074829 | |
3,3'-diethylthiadicarbocyanine iodide | |
(2E)-3-ethyl-2-[(2E,4E)-5-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzothiazole;hydroiodide |
Safety and Handling
GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 208-186-7
RTECSNumber : DL7060000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only