missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3'-Diethyloxacarbocyanine iodide, 98%

$48.86 - $638.65

Chemical Identifiers

CAS 905-96-4
Molecular Formula C21H22IN2O2+
Molecular Weight (g/mol) 461.323
MDL Number MFCD00011952
InChI Key FIZZUEJIOKEFFZ-UHFFFAOYSA-N
Synonym 3,3'-diethyloxacarbocyanine iodide
PubChem CID 129893627
IUPAC Name (2Z)-3-ethyl-2-[(E)-3-(3-ethyl-1,3-benzoxazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzoxazole;hydroiodide
SMILES CCN1C2=CC=CC=C2OC1=CC=CC3=[N+](C4=CC=CC=C4O3)CC.I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55607MD View Documents Alfa Aesar™
H55607MD
250mg Each for $48.86
 
AAH5560703 View Documents Alfa Aesar™
H5560703
1g Each for $144.66
 
AAH5560706 View Documents Alfa Aesar™
H5560706
5g Each for $638.65
 
Specifications

Chemical Identifiers

905-96-4
461.323
FIZZUEJIOKEFFZ-UHFFFAOYSA-N
129893627
CCN1C2=CC=CC=C2OC1=CC=CC3=[N+](C4=CC=CC=C4O3)CC.I
C21H22IN2O2+
MFCD00011952
3,3'-diethyloxacarbocyanine iodide
(2Z)-3-ethyl-2-[(E)-3-(3-ethyl-1,3-benzoxazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzoxazole;hydroiodide

Specifications

Light sensitive
278°C (decomposition)
905-96-4
MFCD00011952
FIZZUEJIOKEFFZ-UHFFFAOYSA-N
(2Z)-3-ethyl-2-[(E)-3-(3-ethyl-1,3-benzoxazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzoxazole;hydroiodide
129893627
Powder
3,3'-Diethyloxacarbocyanine iodide
Red
250mg
C21H22IN2O2+
3,3'-diethyloxacarbocyanine iodide
CCN1C2=CC=CC=C2OC1=CC=CC3=[N+](C4=CC=CC=C4O3)CC.I
461.323
460.31
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 212-996-6

TSCA : Yes

Recommended Storage : Ambient temperatures