missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3'-Diethyl-9-methylthiacarbocyanine iodide, 98%

Manufacturer:  Alfa Aesar™ H5514403

 View more versions of this product

Catalog No. AAH5514403



Specifications

Specifications

3,3'-Diethyl-9-methylthiacarbocyanine iodide
275°C to 285°C
3065-79-0
MFCD00011955
SAEMBGFHGROQJZ-UHFFFAOYSA-N
(2Z)-3-ethyl-2-[(E)-3-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)-2-methylprop-2-enylidene]-1,3-benzothiazole;hydroiodide
129893658
Solid
Light Sensitive
1g
C22H24IN2S2+
3,3'-diethyl-9-methylthiacarbocyanine iodide
CCN1C2=CC=CC=C2SC1=CC(=CC3=[N+](C4=CC=CC=C4S3)CC)C.I
507.472
506.46
98%

Chemical Identifiers

3065-79-0
507.472
SAEMBGFHGROQJZ-UHFFFAOYSA-N
129893658
CCN1C2=CC=CC=C2SC1=CC(=CC3=[N+](C4=CC=CC=C4S3)CC)C.I
C22H24IN2S2+
MFCD00011955
3,3'-diethyl-9-methylthiacarbocyanine iodide
(2Z)-3-ethyl-2-[(E)-3-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)-2-methylprop-2-enylidene]-1,3-benzothiazole;hydroiodide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DL7047000

TSCA : TSCA

Documents