Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3-Diethoxy-trans-1-propenylboronic acid pinacol ester, 97%

$173.28 - $480.66

Chemical Identifiers

Chemical Name or Material 3,3-Diethoxy-trans-1-propenylboronic acid pinacol ester
CAS 153737-25-8
Molecular Formula C13H25BO4
Formula Weight 256.15
MDL Number MFCD03788737
Synonym 2-[(1E)-3,3-Diethoxy-1-propen-1-yl]-4,4,5,5,-tetramethyl-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1957903

  SDS
alfa aesar™
L1957903
1g Each for $480.66

AAL19579MD

  SDS
alfa aesar™
L19579MD
250mg Each for $173.28
Description & Specifications

Specifications

Formula Weight 256.15
Chemical Name or Material 3,3-Diethoxy-trans-1-propenylboronic acid pinacol ester
MDL Number MFCD03788737
Assay 97%
Molecular Formula C13H25BO4
CAS 153737-25-8
Synonym 2-[(1E)-3,3-Diethoxy-1-propen-1-yl]-4,4,5,5,-tetramethyl-1,3,2-dioxaborolane
Quantity 1g

Applications
3,3-Diethoxy-trans-1-propenylboronic acid pinacol ester is used as a synthetic and pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.