missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%, contains up to 10% water, ACROS Organics™

$40.76 - $441.39

Chemical Identifiers

CAS 868272-85-9
Molecular Formula 4HCl·xH2O
Molecular Weight (g/mol) 360.11
MDL Number MFCD08273058
InChI Key WWIHRTPUMJFLTN-UHFFFAOYSA-N
Synonym 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride hydrate, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride xhydrate, 3,3 inverted exclamation marka-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine hydrate tetrahydrochloride, 3,3'-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, for spectrophotometric det. of se, 3,3'-diaminobenzidine tetrahydrochloride hydrate, isopac r, acmc-209qbm, biphenyl-3,3',4,4'-tetramine tetrahydrochloride hydrate
PubChem CID 23191111
IUPAC Name 4-(3,4-diaminophenyl)benzene-1,2-diamine;hydrate
SMILES C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N.O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC112090010
SDS View Documents
Thermo Fisher Scientific
AC112090010
1g Glass bottle
Each for $40.76
 
AC112090050
SDS View Documents
Fisher Scientific Acros
112090050
5g Glass bottle
Each for $93.99
 
AC112090250
SDS View Documents
Fisher Scientific Acros
112090250
25g Glass bottle
Each for $441.39
 
Description

Description

Specifications

Chemical Identifiers

868272-85-9
360.11
WWIHRTPUMJFLTN-UHFFFAOYSA-N
23191111
C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N.O
4HCl·xH2O
MFCD08273058
1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride hydrate, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride xhydrate, 3,3 inverted exclamation marka-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine hydrate tetrahydrochloride, 3,3'-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, for spectrophotometric det. of se, 3,3'-diaminobenzidine tetrahydrochloride hydrate, isopac r, acmc-209qbm, biphenyl-3,3',4,4'-tetramine tetrahydrochloride hydrate
4-(3,4-diaminophenyl)benzene-1,2-diamine;hydrate

Specifications

Authentic
Glass bottle
300.0°C
868272-85-9,868272-85-9
100.0
4HCl·xH2O
MFCD08273058
1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride hydrate, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride xhydrate, 3,3 inverted exclamation marka-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine hydrate tetrahydrochloride, 3,3'-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, for spectrophotometric det. of se, 3,3'-diaminobenzidine tetrahydrochloride hydrate, isopac r, acmc-209qbm, biphenyl-3,3',4,4'-tetramine tetrahydrochloride hydrate
WWIHRTPUMJFLTN-UHFFFAOYSA-N
4-(3,4-diaminophenyl)benzene-1,2-diamine;hydrate
23191111
Crystalline Powder
3, 3'-Diaminobenzidine tetrahydrochloride hydrate
10% max. (120°C)
Brown-Purple to Gray
1g
96.0
96% min. (on dry substance) (Argentometry)
4HCl·xH2O
13, 340
Solubility in water: moderately soluble. Other solubilities: insoluble in most common organic solvents
C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N.O
360.11
360.11
97%
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer.
Suspected of causing genetic defects.

GHS P Statement
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.

GHS Signal Word: Danger

EINECSNumber : 231-018-9

RTECSNumber : DV8753000

TSCA : TSCA

SDS