missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3'-Diaminobenzidine, 99%, ACROS Organics™

$52.82 - $52.82

Chemical Identifiers

CAS 91-95-2
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C12H14N4
Formula Weight 214.27
MDL Number MFCD00007725
Synonym 3,3',4,4'-Biphenyltetramine
Chemical Name or Material 3, 3'-Diaminobenzidine, 99%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC112080050 SDS Fisher Scientific Acros
112080050
Glass bottle 5g Each for $52.82
Specifications Description & Specifications

Chemical Identifiers

91-95-2
100.0
C12H14N4
MFCD00007725
3, 3'-Diaminobenzidine, 99%
98.5
99%
214.27
3,3',4,4'-Biphenyltetramine

Specifications

5g
98.5
Glass bottle
C12H14N4
MFCD00007725
3,3',4,4'-Biphenyltetramine
91-95-2
100.0
99%
214.27
13, 340
3, 3'-Diaminobenzidine, 99%

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.
May cause cancer.

GHS P Statement:
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 202-110-6

RTECSNumber : DV8750000

TSCA : TSCA