missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3'-Diaminobenzidine, 99%

Manufacturer:  Alfa Aesar™ H5400014

 View more versions of this product

Catalog No. AAH5400014



Description

Description

Applications
3,3′-Diaminobenzidine is used as peroxidase substrate and a reagent for spectrophotometric determination of selenium. DAB is used for the immunohistochemical staining of nucleic acids and proteins. As a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue. It may be used for the synthesis of a useful, linear, Schiff-base coordination polymer.

Solubility
Soluble in water (partly).

Notes
Light sensitive. Incompatible with oxidizing agents and heat.
Specifications

Specifications

3,3'-Diaminobenzidine
175°C to 177°C
91-95-2
MFCD00007725
Soluble in water (partly).
C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N
214.272
214.27
Light sensitive
25g
C12H14N4
1,1'-biphenyl-3,3',4,4'-tetramine
HSTOKWSFWGCZMH-UHFFFAOYSA-N
4-(3,4-diaminophenyl)benzene-1,2-diamine
7071
99%

Chemical Identifiers

91-95-2
214.272
HSTOKWSFWGCZMH-UHFFFAOYSA-N
7071
C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N
C12H14N4
MFCD00007725
1,1'-biphenyl-3,3',4,4'-tetramine
4-(3,4-diaminophenyl)benzene-1,2-diamine
Safety and Handling

Safety and Handling

GHS H Statement
H350-H341
May cause cancer.
Suspected of causing genetic defects.

P201-P202-P281-P308+P313-P501c

H341-H350

EINECSNumber : 202-110-6

RTECSNumber : DV8750000

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

Documents