missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3'-Diallylbisphenol A diacetate, epoxy curative, EC-392

Manufacturer:  Alfa Aesar™ 4640314

 View more versions of this product

Catalog No. AA4640314



Description

Description

  • Recommended for use in epoxy systems or in combination with acrylates, methacrylates, and maleimides
Specifications

Specifications

3,3'-Diallylbisphenol A diacetate
1071466-61-9
MFCD22200539
GJTZQXNRNNDGRG-UHFFFAOYSA-N
[4-[2-(4-acetyloxy-3-prop-2-enylphenyl)propan-2-yl]-2-prop-2-enylphenyl] acetate
59215472
Liquid
25g
Epoxy curative, EC-392
C25H28O4
2,2'-diallyl bisphenol a diacetate ether, 4,4'-isopropylidene bis 2-allylphenol acetate, phenyl ester epoxy curative hybrid of bisphenol a, 3,3'-diallylbisphenol a diacetate, epoxy curative, ec-392, 4-2-4-acetyloxy-3-prop-2-en-1-yl phenyl propan-2-yl-2-prop-2-en-1-yl phenyl acetate
CC(=O)OC1=C(C=C(C=C1)C(C)(C)C2=CC(=C(C=C2)OC(=O)C)CC=C)CC=C
392.495
392.5
Amber

Chemical Identifiers

1071466-61-9
392.495
GJTZQXNRNNDGRG-UHFFFAOYSA-N
59215472
CC(=O)OC1=C(C=C(C=C1)C(C)(C)C2=CC(=C(C=C2)OC(=O)C)CC=C)CC=C
C25H28O4
MFCD22200539
2,2'-diallyl bisphenol a diacetate ether, 4,4'-isopropylidene bis 2-allylphenol acetate, phenyl ester epoxy curative hybrid of bisphenol a, 3,3'-diallylbisphenol a diacetate, epoxy curative, ec-392, 4-2-4-acetyloxy-3-prop-2-en-1-yl phenyl propan-2-yl-2-prop-2-en-1-yl phenyl acetate
[4-[2-(4-acetyloxy-3-prop-2-enylphenyl)propan-2-yl]-2-prop-2-enylphenyl] acetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents