Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol, 99%

$40.34 - $40.34

Chemical Identifiers

Chemical Name or Material (+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol
CAS 101203-31-0
Molecular Formula C24H34O2
Formula Weight 354.54
MDL Number MFCD01862439
Synonym BIPHEN-H2; (+/-)-3,3'-Di-tert-butyl-2,2'-dihydroxy-5,5',6,6'-tetramethylbiphenyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2694903

  SDS
alfa aesar™
H2694903
1g Each for $40.34
Description & Specifications

Specifications

Formula Weight 354.54
Chemical Name or Material (+/-)-3,3'-Di-tert-butyl-5,5',6,6'-tetramethylbiphenyl-2,2'-diol
Assay 99%
Molecular Formula C24H34O2
MDL Number MFCD01862439
CAS 101203-31-0
Synonym BIPHEN-H2; (+/-)-3,3'-Di-tert-butyl-2,2'-dihydroxy-5,5',6,6'-tetramethylbiphenyl
Melting Point 164° to 169°C
Quantity 1g