Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3'-Di-n-hexyloxacarbocyanine iodide, 98%

Chemical Identifiers

Chemical Name or Material 3,3'-Di-n-hexyloxacarbocyanine iodide
CAS 53213-82-4
Formula Weight 572.53
MDL Number MFCD00011954
Beilstein 9312562
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5557403

 
alfa aesar™
H5557403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5557406

 
alfa aesar™
H5557406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55574MD

 
alfa aesar™
H55574MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 219° to 221°C
Formula Weight 572.53
Chemical Name or Material 3,3'-Di-n-hexyloxacarbocyanine iodide
TSCA TSCA
Quantity 1g
Color Red
Sensitivity Light Sensitive
Physical Form Powder
Assay 98%
MDL Number MFCD00011954
CAS 53213-82-4
Beilstein 9312562