missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3'-Di-n-hexyloxacarbocyanine iodide, 98%

Manufacturer:  Alfa Aesar™ H55574MD

 View more versions of this product

Catalog No. AAH55574MD



Specifications

Specifications

3,3'-Di-n-hexyloxacarbocyanine iodide
Red
250mg
C29H38IN2O2+
9312562
XVLXYDXJEKLXHN-UHFFFAOYSA-N
(2Z)-3-hexyl-2-[(E)-3-(3-hexyl-1,3-benzoxazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzoxazole;hydroiodide
129893825
Powder
Light Sensitive
219°C to 221°C
53213-82-4
MFCD00011954
3,3'-dihexyloxacarbocyanine iodide
CCCCCCN1C2=CC=CC=C2OC1=CC=CC3=[N+](C4=CC=CC=C4O3)CCCCCC.I
573.539
572.53
98%

Chemical Identifiers

53213-82-4
573.539
XVLXYDXJEKLXHN-UHFFFAOYSA-N
129893825
CCCCCCN1C2=CC=CC=C2OC1=CC=CC3=[N+](C4=CC=CC=C4O3)CCCCCC.I
C29H38IN2O2+
MFCD00011954
3,3'-dihexyloxacarbocyanine iodide
(2Z)-3-hexyl-2-[(E)-3-(3-hexyl-1,3-benzoxazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzoxazole;hydroiodide
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents