missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3'-Di-n-hexyloxacarbocyanine iodide, 98%

Manufacturer:  Alfa Aesar™ H5557406

 View more versions of this product

Catalog No. AAH5557406



Specifications

Specifications

3,3'-Di-n-hexyloxacarbocyanine iodide
Red
5g
C29H38IN2O2+
9312562
XVLXYDXJEKLXHN-UHFFFAOYSA-N
(2Z)-3-hexyl-2-[(E)-3-(3-hexyl-1,3-benzoxazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzoxazole;hydroiodide
129893825
Powder
Light sensitive
219°C to 221°C
53213-82-4
MFCD00011954
3,3'-dihexyloxacarbocyanine iodide
CCCCCCN1C2=CC=CC=C2OC1=CC=CC3=[N+](C4=CC=CC=C4O3)CCCCCC.I
573.539
572.53
98%

Chemical Identifiers

53213-82-4
573.539
XVLXYDXJEKLXHN-UHFFFAOYSA-N
129893825
CCCCCCN1C2=CC=CC=C2OC1=CC=CC3=[N+](C4=CC=CC=C4O3)CCCCCC.I
C29H38IN2O2+
MFCD00011954
3,3'-dihexyloxacarbocyanine iodide
(2Z)-3-hexyl-2-[(E)-3-(3-hexyl-1,3-benzoxazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzoxazole;hydroiodide
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents