Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3'-Di-n-heptyloxacarbocyanine iodide, 97%

Chemical Identifiers

Chemical Name or Material 3,3'-Di-n-heptyloxacarbocyanine iodide
CAS 53213-83-5
Formula Weight 600.58
MDL Number MFCD00274413
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH55777MC

 
alfa aesar™
H55777MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55777ME

 
alfa aesar™
H55777ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 600.58
Sensitivity Light Sensitive
Color Red
Quantity 100mg
Assay 97%
CAS 53213-83-5
Chemical Name or Material 3,3'-Di-n-heptyloxacarbocyanine iodide
MDL Number MFCD00274413
Physical Form Solid