Please login to your online account to display your discounted pricing

3-(3-Chlorophenyl)-3-oxopropanenitrile, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(3-Chlorophenyl)-3-oxopropanenitrile
CAS 21667-62-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H6ClNO
Linear Formula ClC6H4C(O)CH2CN
Formula Weight 179.61
MDL Number MFCD00067891
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM01406EA

 
fisher scientific company
KM01406EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.61
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 3-(3-Chlorophenyl)-3-oxopropanenitrile
Assay Percent Range 97%
Linear Formula ClC6H4C(O)CH2CN
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
MDL Number MFCD00067891
CAS 21667-62-9
Molecular Formula C9H6ClNO