missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3-Chlorophenoxy)piperidine hydrochloride

$94.81 - $341.72

Chemical Identifiers

CAS 1185301-51-2
Molecular Formula C11H14ClNO·HCl
Formula Weight 248.15
MDL Number MFCD11870724
Chemical Name or Material 3-(3-Chlorophenoxy)piperidine hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5118003 SDS Alfa Aesar™
H5118003
1g Each for $341.72
AAH51180MD SDS Alfa Aesar™
H51180MD
250mg Each for $94.81
Specifications Description & Specifications

Chemical Identifiers

1185301-51-2
248.15
3-(3-Chlorophenoxy)piperidine hydrochloride
C11H14ClNO·HCl
MFCD11870724

Specifications

1g
C11H14ClNO·HCl
MFCD11870724
1185301-51-2
248.15
3-(3-Chlorophenoxy)piperidine hydrochloride

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning