missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3'-Carbonylbis(azanediyl)bis(3,1-phenylene)diboronic acid, 97%

Manufacturer:  Alfa Aesar™ H53320MD

 View more versions of this product

Catalog No. AAH53320MD



Specifications

Specifications

3,3'-Carbonylbis(azanediyl)bis(3,1-phenylene)diboronic acid
250mg
C13H14B2N2O5
1,3-bis 3-boronophenyl urea, carbonylbis azanediyl bis 3,1-phenylene diboronic acid, 3-3-dihydroxyboranyl phenyl carbamoyl amino phenylboronic acid, carbonylbis azanediyl-3,1-phenylene diboronic acid
B(C1=CC(=CC=C1)NC(=O)NC2=CC=CC(=C2)B(O)O)(O)O
299.884
299.88
225°C to 226°C
957060-87-6
MFCD09027259
UQUQNYFFKZOTSS-UHFFFAOYSA-N
[3-[(3-boronophenyl)carbamoylamino]phenyl]boronic acid
44119184
97%

Chemical Identifiers

957060-87-6
299.884
UQUQNYFFKZOTSS-UHFFFAOYSA-N
44119184
B(C1=CC(=CC=C1)NC(=O)NC2=CC=CC(=C2)B(O)O)(O)O
C13H14B2N2O5
MFCD09027259
1,3-bis 3-boronophenyl urea, carbonylbis azanediyl bis 3,1-phenylene diboronic acid, 3-3-dihydroxyboranyl phenyl carbamoyl amino phenylboronic acid, carbonylbis azanediyl-3,1-phenylene diboronic acid
[3-[(3-boronophenyl)carbamoylamino]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents