missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(3-Bromophenyl)-1-methyl-1H-pyrazole, 97%, Maybridge

Chemical Identifiers

CAS 425379-68-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H9BrN2
Formula Weight 237.1
MDL Number MFCD09065008
Chemical Name or Material 3-(3-Bromophenyl)-1-methyl-1H-pyrazole
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC60610DA SDS Fisher Scientific Company
CC60610DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC60610DE SDS Fisher Scientific Company
CC60610DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

425379-68-6
100.0
C10H9BrN2
MFCD09065008
97%
96.5
97%
237.1
3-(3-Bromophenyl)-1-methyl-1H-pyrazole

Specifications

425379-68-6
100.0
1g
C10H9BrN2
MFCD09065008
97%
96.5
Amber Glass Bottle
97%
237.1
3-(3-Bromophenyl)-1-methyl-1H-pyrazole

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.