missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(3-Bromophenoxy)-N,N-dimethylpropylamine, 97%, Maybridge

Chemical Identifiers

CAS 912569-57-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H16BrNO
Formula Weight 258.16
MDL Number MFCD03718729
Chemical Name or Material 3-(3-Bromophenoxy)-N, N-dimethylpropylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC59410DA SDS Fisher Scientific Company
CC59410DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC59410DE SDS Fisher Scientific Company
CC59410DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

912569-57-4
100.0
C11H16BrNO
MFCD03718729
97%
96.5
97%
258.16
3-(3-Bromophenoxy)-N, N-dimethylpropylamine

Specifications

258.16
100.0
912569-57-4
C11H16BrNO
Amber Glass Bottle
97%
1g
MFCD03718729
97%
96.5
3-(3-Bromophenoxy)-N, N-dimethylpropylamine

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger