Please login to your online account to display your discounted pricing

3-[3-(Bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole 97+%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC39108DE

 View more versions of this product

Catalog No. CC39108DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 253.1
Quantity 5g
Assay 97%
Chemical Name or Material 3-[3-(Bromomethyl)phenyl]-5-methyl-1, 2, 4-oxadiazole
Assay Percent Range 97%
Molecular Formula C10H9BrN2O
CAS 253273-90-4
Packaging Amber Glass Bottle

Specifications

Chemical Name or Material 3-[3-(Bromomethyl)phenyl]-5-methyl-1, 2, 4-oxadiazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H9BrN2O
Formula Weight 253.1