Please login to your online account to display your discounted pricing

3-[3-(Bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole 97+%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-[3-(Bromomethyl)phenyl]-5-methyl-1, 2, 4-oxadiazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H9BrN2O
Formula Weight 253.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC39108DE

 
fisher scientific company
CC39108DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC39108CB

  SDS
fisher scientific company
CC39108CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC39108DA

  SDS
fisher scientific company
CC39108DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 253.1
Quantity 5g
Assay 97%
Assay Percent Range 97%
Chemical Name or Material 3-[3-(Bromomethyl)phenyl]-5-methyl-1, 2, 4-oxadiazole
CAS 253273-90-4
Molecular Formula C10H9BrN2O
Packaging Amber Glass Bottle