missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3′-Bis(trimethylsilyl)biphenyl-4,4′-diyl Bis(trifluoromethanesulfonate), TCI America™

$1,078.64 - $1,885.31

Chemical Identifiers

CAS 828282-80-0
Molecular Formula C20H24F6O6S2Si2
Molecular Weight (g/mol) 594.686
InChI Key FDAGZCNNWNNCEK-UHFFFAOYSA-N
Synonym 3,3′C-Bis(trimethylsilyl)biphenyl-4,4′C-diyl Ditriflate, 4,4′C-Bis(trifluoromethanesulfonyloxy)-3,3′C-bis(trimethylsilyl)biphenyl
PubChem CID 12182649
IUPAC Name [4-[4-(trifluoromethylsulfonyloxy)-3-trimethylsilylphenyl]-2-trimethylsilylphenyl] trifluoromethanesulfonate
SMILES C[Si](C)(C)C1=C(C=CC(=C1)C2=CC(=C(C=C2)OS(=O)(=O)C(F)(F)F)[Si](C)(C)C)OS(=O)(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B30471G View Documents TCI America
B30471G
1g Each for $1,078.64
 
B30475G View Documents TCI America
B30475G
5g Each for $1,885.31
 
Specifications

Chemical Identifiers

828282-80-0
594.686
3,3′C-Bis(trimethylsilyl)biphenyl-4,4′C-diyl Ditriflate, 4,4′C-Bis(trifluoromethanesulfonyloxy)-3,3′C-bis(trimethylsilyl)biphenyl
[4-[4-(trifluoromethylsulfonyloxy)-3-trimethylsilylphenyl]-2-trimethylsilylphenyl] trifluoromethanesulfonate
C20H24F6O6S2Si2
FDAGZCNNWNNCEK-UHFFFAOYSA-N
12182649
C[Si](C)(C)C1=C(C=CC(=C1)C2=CC(=C(C=C2)OS(=O)(=O)C(F)(F)F)[Si](C)(C)C)OS(=O)(=O)C(F)(F)F

Specifications

White
1g
C20H24F6O6S2Si2
3,3′C-Bis(trimethylsilyl)biphenyl-4,4′C-diyl Ditriflate, 4,4′C-Bis(trifluoromethanesulfonyloxy)-3,3′C-bis(trimethylsilyl)biphenyl
C[Si](C)(C)C1=C(C=CC(=C1)C2=CC(=C(C=C2)OS(=O)(=O)C(F)(F)F)[Si](C)(C)C)OS(=O)(=O)C(F)(F)F
594.686
594.69
≥98.0% (GC)
118°C
828282-80-0
3261
FDAGZCNNWNNCEK-UHFFFAOYSA-N
[4-[4-(trifluoromethylsulfonyloxy)-3-trimethylsilylphenyl]-2-trimethylsilylphenyl] trifluoromethanesulfonate
12182649
Crystal-Powder at 20°C
3,3′-Bis(trimethylsilyl)biphenyl-4,4′-diyl Bis(trifluoromethanesulfonate)
Safety and Handling

Safety and Handling

TSCA : No