missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3-Aminophenyl)propionic acid, 97%

$80.96 - $252.45

Chemical Identifiers

CAS 1664-54-6
Molecular Formula C9H11NO2
Formula Weight 165.19
MDL Number MFCD00153889
Beilstein 2804090
Chemical Name or Material 3-(3-Aminophenyl)propionic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1932903 SDS Alfa Aesar™
L1932903
1g Each for $80.96
AAL1932906 SDS Alfa Aesar™
L1932906
5g Each for $252.45
Specifications

Description

Applications
3-(3-Aminophenyl)propionic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

1664-54-6
165.19
2804090
C9H11NO2
MFCD00153889
3-(3-Aminophenyl)propionic acid

Specifications

1664-54-6
1g
165.19
2804090
97%
99° to 101°C
C9H11NO2
MFCD00153889
3-(3-Aminophenyl)propionic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning