Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3,5-Trimethylcyclohexyl methacrylate, mixture of isomers, 97%, stab. with 200ppm 4-methoxyphenol

Chemical Identifiers

Chemical Name or Material 3,3,5-Trimethylcyclohexyl methacrylate
CAS 7779-31-9
Name Note mixture of isomers
Molecular Formula C13H22O2
Formula Weight 210.32g/mol
MDL Number MFCD00078372
Beilstein 3245062
Synonym Isophoryl methacrylate; Methacrylic acid 3,3,5-trimethylcyclohexyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1219309

 
alfa aesar™
L12193-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1219318

 
alfa aesar™
L12193-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 80° to 81°C (2mmHg)
Quantity 10g
Flash Point 97°C (206°F)
Chemical Name or Material 3,3,5-Trimethylcyclohexyl methacrylate
Name Note mixture of isomers
CAS 7779-31-9
Molecular Formula C13H22O2
Formula Weight 210.32g/mol
MDL Number MFCD00078372
Beilstein 3245062
Synonym Isophoryl methacrylate; Methacrylic acid 3,3,5-trimethylcyclohexyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P305+P351+P338-P302+P352
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P305+P351+P338-P302+P352
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.