missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3,5-Dimethoxyphenyl)propionic acid, 97%

Manufacturer:  Alfa Aesar™ H5400806

 View more versions of this product

Catalog No. AAH5400806



Specifications

Specifications

3-(3,5-Dimethoxyphenyl)propionic acid
5g
C11H14O4
3,5-dimethoxybenzenepropionic acid
COC1=CC(=CC(=C1)CCC(=O)O)OC
210.229
210.23
99°C
717-94-2
MFCD03425685
LMBOJOXVLORKSQ-UHFFFAOYSA-N
3-(3,5-dimethoxyphenyl)propanoic acid
3870216
97%

Chemical Identifiers

717-94-2
210.229
LMBOJOXVLORKSQ-UHFFFAOYSA-N
3870216
COC1=CC(=CC(=C1)CCC(=O)O)OC
C11H14O4
MFCD03425685
3,5-dimethoxybenzenepropionic acid
3-(3,5-dimethoxyphenyl)propanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents